Learn More
American Research Products Inc Mouse Dickkopf related protein
Catalog No. 5000327222
Encompass
Mouse Dickkopf-related protein 3 (DKK3) ELISA kit from CUSABIO Detection Range 9 38 pg/ml-600 pg/ml Sensitivity 2 34 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27222 Supplier American Research Products Inc Supplier No. CSBEL006922MO
Mouse Dickkopf-related protein 3 (DKK3) ELISA kit from CUSABIO Detection Range 9 38 pg/ml-600 pg/ml Sensitivity 2 34 pg/ml Sample serum plasma tissue homogenates Method Sandwich