Learn More
American Research Products Inc Mouse Dickkopf related protein
Supplier:  American Research Products Inc CSBEL006920MO
Encompass
Mouse Dickkopf-related protein 1 (DKK1) ELISA kit from CUSABIO Detection Range 78 125 pg/mL-5000 pg/mL Sensitivity 13 127 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28048