Learn More
Biomatik Corporation Mouse Dihydropyrimidinase Like Protein 2 (DPYSL2) ELISA Kit
Supplier:  Biomatik Corporation EKU10794
Encompass_Preferred
Mouse Dihydropyrimidinase Like Protein 2 (DPYSL2) ELISA Kit for detecting DPYSL2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08553. Gene ID: 12934. Target Synonyms: CRMP2; DHPRP2; DRP-2; DRP2; ULIP2; Unc-33-like phosphoprotein 2; Collapsin response mediator protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5380