Learn More
Biomatik Corporation Mouse Dihydropyrimidine Dehydrogenase (DPYD) ELISA Kit
Supplier:  Biomatik Corporation EKU03742
Encompass_Preferred
Mouse Dihydropyrimidine Dehydrogenase (DPYD) ELISA Kit for detecting DPYD in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8CHR6. Gene ID: 99586. Target Synonyms: DPD; DHPDHase; Dihydrothymine dehydrogenase; Dihydrouracil dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7368