Learn More
American Research Products Inc Mouse dipeptidyl peptldase IV
Catalog No. 5000330228
Encompass
Mouse dipeptidyl peptldase IV DPPIV ELISA Kit from CUSABIO Detection Range 0 312 ng/ml-20 ng/ml Sensitivity 0 078 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30228 Supplier American Research Products Inc Supplier No. CSBE08520M
Mouse dipeptidyl peptldase IV DPPIV ELISA Kit from CUSABIO Detection Range 0 312 ng/ml-20 ng/ml Sensitivity 0 078 ng/ml Sample serum plasma tissue homogenates Method Sandwich