Learn More
Selleck Chemical LLC Mouse Direct PCR Kit
SDP
Catalog No. 503134722
Encompass_Preferred
The Mouse Direct PCR Kit provides a fast preparation and PCR amplIFication that is specIFically designed for mouse genotyping Buffer L and Protease Plus rapidly digests mouse tissue to release intact genomic DNA that can be used directly as the template for PCR amplIFication
Catalog No. 50-313-4722 Supplier Selleck Chemical LLC Supplier No. B40013200RXNS
The Mouse Direct PCR Kit provides a fast preparation and PCR amplIFication that is specIFically designed for mouse genotyping Buffer L and Protease Plus rapidly digests mouse tissue to release intact genomic DNA that can be used directly as the template for PCR amplIFication