Learn More
Sino Biological Recombinant Mouse DKK-1 Protein (His Tag)
Supplier:  Sino Biological 57248M08H1
Encompass_Preferred
A DNA sequence encoding the mouse DKK1 (NP_034181.2) (Met1-His272) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2954