Learn More
Sino Biological Anti-DKK-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50247RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Dkk-3 / Dickkopf-3 (rM Dkk3; Catalog#50247-M08H; NP_056629.1; Met 1-Ile 349). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3340