Learn More
Sino Biological Anti-DLL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50522RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse DLL1 (rM DLL1; Catalog#50522-M08H; NP_031891.2; Met 1-Gln 516). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2134