Learn More
ACROBiosystems Mouse DLL3 Protein, His Tag (Ala 25-Arg 488), HEK293-expressed, lyophilized, 100 μg
Supplier:  ACROBiosystems DL3M52H9100UG
Encompass_Preferred
Recombinant extracellular domain of mouse Delta-like protein 3 (Ala 25-Arg 488) expressed in HEK293 cells, C-terminal His-tagged, lyophilized and formulated for use in antibody characterization and binding assays.
  • Expressed in HEK293 cells for mammalian glycosylation and folding.
  • C-terminal polyhistidine tag with an N-terminal fusion design.
  • Purity greater than 95% by SDS-PAGE and >85% by SEC-HPLC.
  • Calculated molecular weight ~118 kDa; migrates ~100-115 kDa by SDS-PAGE.
  • Endotoxin less than 1.0 EU per μg (LAL/rFC).
  • Assay verification includes SDS-PAGE, SEC-HPLC, bioactivity ELISA, and bioactivity SPR.
  • Supplied lyophilized from PBS, pH 7.4, with trehalose; reconstitute per manufacturer instructions.
  • Intended for antibody binding assays, ELISA, SPR, and assay development.
Catalog No. NC2121668