Learn More
Sino Biological Recombinant Anti-DLL4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50640R00420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DLL4 / Delta-like 4 (rM DLL4 / Delta-like 4; Catalog#50640-M08H; NP_062327.2; Met1-Pro525).
Catalog No. 50-241-0713