Learn More
Sino Biological Anti-DLL4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50640RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse DLL4 (rM DLL4; Catalog#50640-M08H; NP_062327.2; Met 1-Pro 525). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1683