Learn More
Sino Biological Recombinant Mouse DLL4 Protein (ECD, mFc Tag)
Supplier:  Sino Biological 50640M38H200
Encompass_Preferred
A DNA sequence encoding the mouse PVR (NP_062327.2) (Met1-Pro525) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-3182