Learn More
Biomatik Corporation Mouse DNA Methyltransferase 3B (DNMT3B) ELISA Kit
Supplier:  Biomatik Corporation EKU03772
Encompass_Preferred
Mouse DNA Methyltransferase 3B (DNMT3B) ELISA Kit for detecting DNMT3B in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.045ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88509. Gene ID: 13436. Target Synonyms: ICF; M.HsaIIIB; DNA methyltransferase HsaIIIB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5795