Learn More
ACROBiosystems IsotypeMouse IgG1/kappaSpecificitySpecifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.ApplicationFlow Cytometry (Used as an Isotype Control for FM3-FY45 and FM3-FY45P1).
Catalog No. 502231537
Encompass_Preferred
Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies. FITC-Labeled Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control was purified from HEK293 cell culture. It is the FITC labeled form of Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control (Cat. No. DNP-M1).
This item is not returnable. View return policy
Catalog No. 50-223-1537 Supplier ACROBiosystems Supplier No. DNPFM1A125TESTS
Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies. FITC-Labeled Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control was purified from HEK293 cell culture. It is the FITC labeled form of Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control (Cat. No. DNP-M1).