Learn More
Sino Biological Recombinant Mouse DPP-4/CD26/DPPIV Protein (hFc Tag)
Supplier:  Sino Biological 50718M01H50
Encompass_Preferred
A DNA sequence encoding the mouse Dpp4 (NP_034204.1) (Ser29-His760) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-224-7098