Learn More
Sino Biological Recombinant Anti-DPP-4/CD26/DPPIV Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50718R009P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DPP4 / CD26 (rM DPP4 / CD26; Catalog#50718-M08H; NP_034204.1; Ser29-His760) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8534