Learn More
Sino Biological Recombinant Mouse Desmocollin 2/DSC2 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 5A0431M08H100
Encompass_Preferred
A DNA sequence encoding the Mouse DSC2 (P55292) (Met1-Pro694) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2942