Learn More
Raybiotech Inc Mouse Dtk ELISA Kit
Supplier:  Raybiotech Inc ELMDTK1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.4 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- ?, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 ?, IL-1?, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1?, MIP-1?, MIP-2, MIP-3?, MIP-3?, PF-4, P-Selectin, RANTES, SCF, SDF-1?, TARC, TCA-3, TECK, TIMP-1, TNF-?, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
Catalog No. 50-194-9968