Learn More
Biomatik Corporation Mouse Dystrophia Myotonica Protein Kinase (DMPK) ELISA Kit
Supplier:  Biomatik Corporation EKU12457
Encompass_Preferred
Mouse Dystrophia Myotonica Protein Kinase (DMPK) ELISA Kit for detecting DMPK in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P54265. Gene ID: 13400. Target Synonyms: DM; DM1; DM1PK; DMK; MDPK; MT-PK; Myotonic Dystrophy Protein Kinase; Myotonin-Protein Kinase; Dystrophia Myotonica 1; Thymopoietin Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6239