Learn More
Sino Biological Anti-E2f6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106121T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse E2F6 , and purified by antigen affinity chromatography.
Catalog No. 50-240-9917