Learn More
American Research Products Inc Mouse E3 ubiquitin protein lig
Catalog No. 5000327171
Encompass
Mouse E3 ubiquitin-protein ligase SMURF1 (SMURF1) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27171 Supplier American Research Products Inc Supplier No. CSBEL021860MO
Mouse E3 ubiquitin-protein ligase SMURF1 (SMURF1) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich