Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Monoclonal Anti-Monkeypox E8L Antibody (Mouse; clone 1C12) - Human IgG1 chimera, 100 μg

Catalog No. 502370254
Encompass_Preferred

Monoclonal anti-Monkeypox E8L antibody (clone 1C12) is a chimeric mouse-derived antibody with human IgG1 constant regions, expressed in HEK293 cells and supplied as an unconjugated preparation for research use.

  • Target: Monkeypox virus E8L protein.
  • Format: Unconjugated antibody, powder or lyophilized.
  • Clone: 1C12.
  • Host: Mouse (chimeric to human IgG1 constant regions).
  • Expression system: HEK293 cells.
  • Purity: Greater than 90%.
  • Size: 100 μg.
  • Storage: Store at -20°C; supplied in PBS, pH 7.4.

Catalog No. 50-237-0254 Supplier ACROBiosystems Supplier No. E8LM573100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monoclonal anti-Monkeypox E8L antibody (clone 1C12) is a chimeric mouse-derived antibody with human IgG1 constant regions, expressed in HEK293 cells and supplied as an unconjugated preparation for research use.

  • Target: Monkeypox virus E8L protein.
  • Format: Unconjugated antibody, powder or lyophilized.
  • Clone: 1C12.
  • Host: Mouse (chimeric to human IgG1 constant regions).
  • Expression system: HEK293 cells.
  • Purity: Greater than 90%.
  • Size: 100 μg.
  • Storage: Store at -20°C; supplied in PBS, pH 7.4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.