Learn More
World Precision Instrument Mouse Ear Bars, Non-Rupture
SDP
Catalog No. 502539185
Encompass
  • These ear bars are specifically designed for use with the Premium Stereotaxic Frames.
  • Ear Bars have laser-engraved scales, 50mm of calibration and are 6.2 mm square. The non-puncture tips have a wider taper than standard ear bars to prevent damage to the ear during placement.
Catalog No. 50-253-9185 Supplier World Precision Instrument Supplier No. 505229
  • These ear bars are specifically designed for use with the Premium Stereotaxic Frames.
  • Ear Bars have laser-engraved scales, 50mm of calibration and are 6.2 mm square. The non-puncture tips have a wider taper than standard ear bars to prevent damage to the ear during placement.