Learn More
BIOHIPPO Mouse ECM1 Microsample
SDP
Catalog No. 5000315940
Encompass
Mouse ECM1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ECM1 in Serum plasma and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 14 72 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse ECM1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse ECM1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse ECM1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15940 Supplier BIOHIPPO Supplier No. BHE15211039
Mouse ECM1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ECM1 in Serum plasma and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 14 72 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse ECM1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse ECM1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse ECM1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color