Learn More
Sino Biological Anti-Ephrin-A1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50593RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Ephrin-A1 / EFNA1 (rM Ephrin-A1 / EFNA1; Catalog#50593-M08H;NP_034237.3; Met 1-Ser 182). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8902