Learn More
Sino Biological Anti-Ephrin A3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50594T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Ephrin-A3 / EFNA3 (Catalog#50594-M08H; NP_034238.1; Met1-Ser205). Ephrin-A3 / EFNA3 specific IgG was purified by Mouse Ephrin-A3 / EFNA3 affinity chromatography.
Catalog No. 50-255-3194