Learn More
Sino Biological Anti-Ephrin-B1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50580RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Ephrin-B1 / EFNB1 (rM Ephrin-B1 / EFNB1; Catalog#50580-M08H; NP_034240.1; Met 1-Ser 229). Ephrin-B1 / EFNB1 specific IgG was purified by Mouse Ephrin-B1 / EFNB1 affinity chromatography.
Catalog No. 50-255-3208