Learn More
Sino Biological Anti-Ephrin B2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50598RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse EFNB2 (rM EFNB2; Catalog#50598-M08H; NP_034241.2; Met 1-Ala 232). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0141