Learn More
Sino Biological Anti-EGF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50482T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse EGF / Epidermal Growth Factor (Catalog#50482-M08H1; AAH60741.1; Met1-Arg1029). EGF / Epidermal Growth Factor specific IgG was purified by Mouse EGF / Epidermal Growth Factor affinity chromatography.
Catalog No. 50-240-8527