Learn More
Sino Biological Recombinant Mouse EGF Protein
Supplier:  Sino Biological 50482MNCH100
Encompass_Preferred
A DNA sequence encoding the mouse EGF (P01132) (Asn977-Arg1029) was expressed and purified with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-255-3256