Learn More
FUJIFILM Biosciences Mouse EGF, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Catalog No. 50900530
Encompass_Preferred
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
Catalog No. 50-900-530 Supplier FUJIFILM Biosciences Supplier No. 200531MG
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).