Learn More
FUJIFILM BIOSCIENCES INC Mouse EGF, Recombinant Protein, Shenandoah Biotechnology, 500 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 20053500UG
Encompass_Preferred
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
Catalog No. 50-900-529