Learn More
Sino Biological Anti-EGF-L6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51182T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse EGFL6 / EGF-L6 (rM EGFL6 / EGF-L6; Catalog#51182-M08H; NP_062270.1; Thr287-Gly550). EGFL6 / EGF-L6 specific IgG was purified by Mouse EGFL6 / EGF-L6 affinity chromatography.
Catalog No. 50-255-2970