Learn More
BIOHIPPO Mouse EGFL7 Microsample
SDP
Catalog No. 5000315821
Encompass
Mouse EGFL7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EGFL7 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EGFL7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EGFL7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EGFL7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15821 Supplier BIOHIPPO Supplier No. BHE15210739
Mouse EGFL7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EGFL7 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EGFL7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EGFL7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EGFL7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color