Learn More
Sino Biological Anti-EGFL7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50472RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse EGFL7 / VE-statin (Catalog#50472-M07B; Q9QXT5-1; Glu22-Leu275). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3263