Learn More
Sino Biological Recombinant Mouse EGFR Protein (hFc Tag) 1
Supplier:  Sino Biological 51091M02H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse EGFR (Q01279) (Met 1-Ser 647) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-7061