Learn More
Sino Biological Recombinant Anti-EGFR Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51091R112100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EGFR / ErbB1 / HER1 (rM EGFR / ErbB1 / HER1; Catalog#51091-M08H; Q01279; Met1-Ser647).
Catalog No. 50-241-1787