Learn More
BIOHIPPO Mouse EGFR Microsample
SDP
Catalog No. 5000314783
Encompass
Mouse EGFR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EGFR in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 93 75-6000 pg/mL with a sensitivity of 38 25 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EGFR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EGFR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EGFR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14783 Supplier BIOHIPPO Supplier No. BHE15212024
Mouse EGFR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EGFR in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 93 75-6000 pg/mL with a sensitivity of 38 25 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EGFR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EGFR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EGFR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color