Learn More
American Research Products Inc Mouse Elastin ELN ELISA kit
Catalog No. 5000328083
Encompass
Mouse Elastin (ELN) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28083 Supplier American Research Products Inc Supplier No. CSBEL007617MO
Mouse Elastin (ELN) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich