Learn More
Biomatik Corporation Mouse Elastin Microfibril Interface Located Protein 2 (EMILIN2) ELISA Kit
Supplier:  Biomatik Corporation EKU03857
Encompass_Preferred
Mouse Elastin Microfibril Interface Located Protein 2 (EMILIN2) ELISA Kit for detecting EMILIN2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.12-5000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K482. Gene ID: 246707. Target Synonyms: FOAP-10; Elastin Microfibril Interfacer 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6049