Learn More
BIOHIPPO Mouse EMILIN2 Microsample
SDP
Catalog No. 5000315394
Encompass
Mouse EMILIN2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EMILIN2 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 27 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EMILIN2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EMILIN2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EMILIN2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15394 Supplier BIOHIPPO Supplier No. BHE15211055
Mouse EMILIN2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EMILIN2 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 27 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse EMILIN2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse EMILIN2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse EMILIN2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color