Learn More
BIOHIPPO Mouse ENA78 Microsample
SDP
Catalog No. 5000313829
Encompass
Mouse ENA78 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ENA78 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 24 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse ENA78 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse ENA78 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse ENA78 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13829 Supplier BIOHIPPO Supplier No. BHE15211954
Mouse ENA78 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ENA78 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 24 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse ENA78 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse ENA78 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse ENA78 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color