Learn More
Biomatik Corporation Mouse Enah/Vasp Like Protein (EVL) ELISA Kit
Supplier:  Biomatik Corporation EKU10805
Encompass_Preferred
Mouse Enah/Vasp Like Protein (EVL) ELISA Kit for detecting EVL in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70429. Target Synonyms: RNB6; Enabled/Vasodilator Stimulated Phosphoprotein Like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8285