Learn More
Raybiotech Inc Mouse Endocan ELISA Kit
Supplier:  Raybiotech Inc ELMENDOCAN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects mouse Endocan, it shows no significant cross-reactivity with recombinant human Endocan.
Catalog No. 50-194-9973