Learn More
Raybiotech Inc Mouse Endostatin ELISA Kit
Supplier:  Raybiotech Inc ELMENDOSTATIN1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 2.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Mouse Endostatin (COL18A1)
Catalog No. 50-194-9974