Learn More
American Research Products Inc Mouse Endothelial nitric oXide
Catalog No. 5000329443
Encompass
Mouse Endothelial nitric oXide synthase eNOS ELISA Kit from CUSABIO Detection Range 0 312 IU/mL-20 IU/mL Sensitivity 0 078 IU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29443 Supplier American Research Products Inc Supplier No. CSBE08324M
Mouse Endothelial nitric oXide synthase eNOS ELISA Kit from CUSABIO Detection Range 0 312 IU/mL-20 IU/mL Sensitivity 0 078 IU/mL Sample serum plasma tissue homogenates Method Sandwich