Learn More
Biomatik Corporation Mouse Enhancer Of Zeste Homolog 2 (EZH2) ELISA Kit
Supplier:  Biomatik Corporation EKU12249
Encompass_Preferred
Mouse Enhancer Of Zeste Homolog 2 (EZH2) ELISA Kit for detecting EZH2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61188. Gene ID: 14056. Target Synonyms: ENX-1; KMT6; KMT6A; Histone-lysine N-methyltransferase EZH2; Lysine N-methyltransferase 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7995