Learn More
Sino Biological Anti-Autotaxin/ENPP-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50663T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ENPP2 (rM ENPP2; Catalog#50663-M07H; Q9R1E6-1; Ser49-Ile862). ENPP2 specific IgG was purified by Mouse ENPP2 affinity chromatography.
Catalog No. 50-255-3158