Learn More
ACROBiosystems ACROBiosystems Mouse ENPP3 / CD203c Protein, His Tag (active enzyme, MALS verified)
Catalog No. 502107275
Encompass_Preferred
Recombinant Protein;100UG;Mouse ENPP3, His Tag (EN3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 46 - Ile 874 (Accession # Q6DYE8-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 95.8 kDa. The protein migrates as 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7275 Supplier ACROBiosystems Supplier No. EN3M52H3100UG
Recombinant Protein;100UG;Mouse ENPP3, His Tag (EN3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 46 - Ile 874 (Accession # Q6DYE8-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 95.8 kDa. The protein migrates as 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.